Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075159.1 | NP_001068627.1 | 7 - 8 | 859 | 92 | |
GenBank mRNA | BC123701.1 | - | 7 - 8 | 859 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |