Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075221.2 | NP_001068689.1 | 1 - 2 | 99 | 116 | |
GenBank mRNA | BC118463.1 | - | 1 - 2 | 101 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |