Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANP32E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075306.1 | NP_001068774.1 | 5 - 6 | 964 | 142 | |
GenBank mRNA | BC120174.1 | - | 5 - 6 | 964 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |