Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075331.1 | NP_001068799.1 | 1 - 2 | 332 | 55 | |
GenBank mRNA | BC120321.1 | - | 1 - 2 | 332 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |