Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WSB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075357.1 | NP_001068825.1 | 6 - 7 | 999 | 67 | |
GenBank mRNA | BC149531.1 | - | 6 - 7 | 953 | 67 | |
BT026227.1 | - | 6 - 7 | 999 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |