Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075477.2 | NP_001068945.1 | 5 - 6 | 668 | 58 | |
GenBank mRNA | BC122807.1 | - | 5 - 6 | 669 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |