Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SURF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075529.1 | NP_001068997.1 | 3 - 4 | 494 | 61 | |
GenBank mRNA | BC119935.1 | - | 3 - 4 | 494 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |