Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075568.1 | NP_001069036.1 | 4 - 5 | 503 | 59 | |
GenBank mRNA | BC119910.1 | - | 4 - 5 | 503 | 59 | |
BT029851.1 | - | 4 - 5 | 451 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |