Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075570.1 | NP_001069038.1 | 9 - 10 | 1280 | 63 | |
GenBank mRNA | BT026545.1 | - | 9 - 10 | 1280 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Inventoried Probe spans exons |