Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STMN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075580.2 | NP_001069048.1 | 2 - 3 | 131 | 70 | |
GenBank mRNA | BC120123.1 | - | 2 - 3 | 132 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |