Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075672.2 | NP_001069140.1 | 2 - 3 | 104 | 78 | |
GenBank mRNA | BC120154.1 | - | 2 - 3 | 105 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |