Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRPEL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075674.1 | NP_001069142.1 | 1 - 2 | 92 | 79 | |
GenBank mRNA | BC119818.1 | - | 1 - 2 | 92 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |