Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAPLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075684.1 | NP_001069152.1 | 4 - 5 | 614 | 56 | |
GenBank mRNA | BC119891.1 | - | 4 - 5 | 614 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |