Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAPLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075684.1 | NP_001069152.1 | 6 - 7 | 913 | 56 | |
GenBank mRNA | BC119891.1 | - | 6 - 7 | 913 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |