Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAXBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075689.3 | NP_001069157.3 | 10 - 11 | 1742 | 158 | |
GenBank mRNA | BC120358.1 | - | 4 - 5 | 681 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |