Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075734.1 | NP_001069202.1 | 2 - 3 | 549 | 67 | |
GenBank mRNA | BC120284.1 | - | 2 - 3 | 549 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |