Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSUN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076001.2 | NP_001069469.1 | 3 - 4 | 486 | 88 | |
GenBank mRNA | BC120181.1 | - | 3 - 4 | 487 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |