Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076042.1 | NP_001069510.1 | 2 - 3 | 2563 | 132 | |
GenBank mRNA | BC126508.1 | - | 3 - 4 | 2575 | 132 | |
BT026193.1 | - | 2 - 3 | 2563 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |