Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PVALB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076114.2 | NP_001069582.1 | 386 | 99 | ||
GenBank mRNA | BC120184.1 | - | 387 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus |