Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MXD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076256.1 | NP_001069724.1 | 5 - 6 | 621 | 154 | |
GenBank mRNA | BC120230.1 | - | 5 - 6 | 621 | 154 | |
BT030600.2 | - | 5 - 6 | 515 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |