Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076296.1 | NP_001069764.1 | 1 - 2 | 171 | 97 | |
GenBank mRNA | BC119926.1 | - | 1 - 2 | 171 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |