Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076316.1 | NP_001069784.1 | 9 - 10 | 1694 | 82 | |
GenBank mRNA | BC122597.1 | - | 9 - 10 | 1694 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |