Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076485.1 | NP_001069953.1 | 1 - 2 | 68 | 102 | |
GenBank mRNA | BC119897.1 | - | 1 - 2 | 68 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |