Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076832.1 | NP_001070300.1 | 4 - 5 | 719 | 96 | |
GenBank mRNA | BC123749.1 | - | 4 - 5 | 719 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |