Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076895.1 | NP_001070363.1 | 4 - 5 | 487 | 69 | |
GenBank mRNA | BC123872.1 | - | 4 - 5 | 487 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |