Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076908.2 | NP_001070376.1 | 13 - 14 | 1923 | 76 | |
GenBank mRNA | BC123530.1 | - | 13 - 14 | 1924 | 76 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |