Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076924.1 | NP_001070392.1 | 13 - 14 | 1695 | 85 | |
GenBank mRNA | BC123459.1 | - | 13 - 14 | 1695 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |