Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076924.1 | NP_001070392.1 | 9 - 10 | 1204 | 76 | |
GenBank mRNA | BC123459.1 | - | 9 - 10 | 1204 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |