Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076928.1 | NP_001070396.1 | 1 - 2 | 644 | 113 | |
GenBank mRNA | DQ922948.1 | - | 1 - 2 | 644 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Bos taurus Probe spans exons |