Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077838.2 | NP_001071306.2 | 1 - 2 | 31 | 74 | |
GenBank mRNA | BC123546.1 | - | 1 - 2 | 32 | 74 | |
EV634454.1 | - | 1 - 2 | 34 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |