Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079614.2 | NP_001073082.1 | 1 - 2 | 386 | 84 | |
GenBank mRNA | BC120248.1 | - | 1 - 2 | 387 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |