Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIF3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080249.2 | NP_001073718.1 | 4 - 5 | 1417 | 72 | |
GenBank mRNA | BC122596.1 | - | 4 - 5 | 1419 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |