Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARRDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080365.2 | NP_001073834.1 | 6 - 7 | 840 | 74 | |
GenBank mRNA | BC120279.1 | - | 6 - 7 | 841 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |