Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080371.2 | NP_001073840.2 | 3 - 4 | 367 | 121 | |
GenBank mRNA | BT026536.1 | - | 2 - 3 | 162 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |