Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083672.2 | NP_001077141.1 | 1 - 2 | 107 | 128 | |
GenBank mRNA | BC122805.1 | - | 1 - 2 | 108 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |