Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173893.3 | NP_776318.1 | 3 - 4 | 409 | 101 | |
GenBank mRNA | AB098926.1 | - | 2 - 3 | 297 | 100 | |
BC118352.1 | - | 3 - 4 | 409 | 101 |
Gene Symbol: LOC783680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB098890.1 | - | 3 - 4 | 392 | 101 | |
X69084.1 | - | 382 | 101 |
Set Membership: |
Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |