Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174447.4 | NP_776872.1 | 4 - 5 | 561 | 77 | |
GenBank mRNA | BC122723.1 | - | 4 - 5 | 562 | 77 | |
M19886.1 | - | 4 - 5 | 517 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |