Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177484.3 | NP_803450.2 | 3 - 4 | 430 | 59 | |
GenBank mRNA | BC122602.1 | - | 3 - 4 | 430 | 59 | |
U02564.1 | - | 3 - 4 | 377 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |