Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TKFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024524.3 | NP_001019695.2 | 4 - 5 | 693 | 77 | |
GenBank mRNA | BC126551.1 | - | 4 - 5 | 694 | 77 | |
BT021593.1 | - | 4 - 5 | 700 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |