Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024561.2 | NP_001019732.1 | 8 - 9 | 1033 | 89 | |
GenBank mRNA | BC133593.1 | - | 8 - 9 | 1033 | 89 | |
BT020721.1 | - | 7 - 8 | 939 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |