Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPADH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025338.2 | NP_001020509.2 | 2 - 3 | 102 | 67 | |
GenBank mRNA | AJ853856.2 | - | 2 - 3 | 102 | 67 | |
AJ853857.2 | - | 2 - 3 | 102 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |