Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIGD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077861.2 | NP_001071329.1 | 1 - 2 | 169 | 82 | |
GenBank mRNA | BC123383.1 | - | 1 - 2 | 170 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |