Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC51B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077867.2 | NP_001071335.1 | 3 - 4 | 355 | 75 | |
GenBank mRNA | BC126778.1 | - | 3 - 4 | 356 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |