Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077868.1 | NP_001071336.1 | 7 - 8 | 790 | 104 | |
GenBank mRNA | BC126666.1 | - | 7 - 8 | 790 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |