Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHEDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077892.2 | NP_001071360.1 | 4 - 5 | 485 | 156 | |
GenBank mRNA | BC126670.1 | - | 4 - 5 | 486 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |