Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4H7orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079785.1 | NP_001073253.1 | 2 - 3 | 226 | 115 | |
GenBank mRNA | BC126755.1 | - | 2 - 3 | 226 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |