Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFEMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081717.2 | NP_001075186.1 | 2 - 3 | 177 | 116 | |
GenBank mRNA | BC133561.1 | - | 2 - 3 | 179 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |