Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082455.2 | NP_001075924.1 | 3 - 4 | 340 | 74 | |
GenBank mRNA | BC133553.1 | - | 3 - 4 | 341 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |