Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC93A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082457.1 | NP_001075926.1 | 5 - 6 | 917 | 67 | |
GenBank mRNA | BC133579.1 | - | 5 - 6 | 917 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |