Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082610.1 | NP_001076079.1 | 1 - 2 | 117 | 67 | |
GenBank mRNA | AY957404.1 | - | 1 - 2 | 117 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |